Learn More
Cole-Parmer 316 Stainless Steel Three-blade Propeller, 1" dia x 5/16" bore dia
Catalog No. NC0217346 Shop All Cole-Parmer Products
Encompass_Preferred
316 Stainless Steel Three-blade Propeller, 1" dia x 5/16" bore diaUpdate clamps, mounts, shafts and propellers to accommodate new protocols. Replacement parts keep your overhead mixer running smoothly.Three-blade propellers are for general-purpose mixing. Use alone or mount several on a single shaft for thorough agitation.
Catalog No. NC0217346 Supplier Cole-Parmer Supplier No. 0455240
316 Stainless Steel Three-blade Propeller, 1" dia x 5/16" bore diaUpdate clamps, mounts, shafts and propellers to accommodate new protocols. Replacement parts keep your overhead mixer running smoothly.Three-blade propellers are for general-purpose mixing. Use alone or mount several on a single shaft for thorough agitation.