Learn More
Cayman Chemical 1-Isothiocyanato-6-(methylsulfenyl)-hexane, Purity: ≥98%, CAS: 4430-39-1, 10 mg
SDP
Catalog No. 502913666
Encompass_Preferred
An isothiocyanate; active against B. subtilis in an agar diffusion assay at 25 µmol/disc and T. mentagrophytes (MIC = 25 µg/ml); has repellent activity in the blue mussel (M. edulis) assay, as well as antifouling activity in the conventional submerged assay when applied to PVC plates at 50 µmol/cm2
Catalog No. 50-291-3666 Supplier Cayman Chemical Supplier No. 3559510 MG
An isothiocyanate; active against B. subtilis in an agar diffusion assay at 25 µmol/disc and T. mentagrophytes (MIC = 25 µg/ml); has repellent activity in the blue mussel (M. edulis) assay, as well as antifouling activity in the conventional submerged assay when applied to PVC plates at 50 µmol/cm2