Learn More
LAMDA BIOTECH INC 1Kb DNA Ladder
Catalog No. 502045811
Encompass_Preferred
1Kb DNA Ladder is a proprietary mixture of several plasmids digested to completion with specific restriction enzymes to produce a ladder with 11 fragments ranging from 0.5-10.0Kb, producing the most complete and convenient ladder in the 1Kb DNA ladder family. The intensity of the some bands has been increased to yield internal reference indicators. 6µl will yield at least of 30ng DNA in any single DNA band.
Catalog No. 50-204-5811 Supplier LAMDA BIOTECH INC Supplier No. M108
1Kb DNA Ladder is a proprietary mixture of several plasmids digested to completion with specific restriction enzymes to produce a ladder with 11 fragments ranging from 0.5-10.0Kb, producing the most complete and convenient ladder in the 1Kb DNA ladder family. The intensity of the some bands has been increased to yield internal reference indicators. 6µl will yield at least of 30ng DNA in any single DNA band.