Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies 1 kb Plus DNA Ladder, 100 μL, 50 ng/μL, Liquid, -20 °C storage, Research Use Only

Catalog No. NC2612435
Encompass

The Agilent 1 kb Plus DNA Ladder is a high-quality molecular weight marker designed for precise sizing of double-stranded DNA fragments. This reliable product is optimized for use with Agilent's Fragment Analyzer and ZAG DNA Analyzer systems, making it an essential tool in genomic and molecular biology applications.

  • Liquid format
  • Concentration of 50 ng/μL
  • Volume of 0.1 mL
  • Sizing range of 75-20000 bp
  • System specific sizing range of 200-10000 bp
  • Storage temperature of -20 °C
  • Guaranteed shelf life of 4 months
  • Compatible with Agilent Fragment Analyzer and ZAG DNA Analyzer systems
  • Kit compatibility with DNF-930 and ZAG-130 kits
  • Application for DNA fragment sizing
  • Regulatory status: Research Use Only (RUO)

Catalog No. NC2612435 Supplier Agilent Technologies Supplier No. FSSLR930U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Agilent 1 kb Plus DNA Ladder is a high-quality molecular weight marker designed for precise sizing of double-stranded DNA fragments. This reliable product is optimized for use with Agilent's Fragment Analyzer and ZAG DNA Analyzer systems, making it an essential tool in genomic and molecular biology applications.

  • Liquid format
  • Concentration of 50 ng/μL
  • Volume of 0.1 mL
  • Sizing range of 75-20000 bp
  • System specific sizing range of 200-10000 bp
  • Storage temperature of -20 °C
  • Guaranteed shelf life of 4 months
  • Compatible with Agilent Fragment Analyzer and ZAG DNA Analyzer systems
  • Kit compatibility with DNF-930 and ZAG-130 kits
  • Application for DNA fragment sizing
  • Regulatory status: Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.