Learn More
LAMDA BIOTECH INC 1Kb Plus DNA Ladder
Catalog No. NC3726411
Encompass_Preferred
The 1Kb Plus™ DNA Ladder-20 is a comprehensive molecular weight marker containing 14 precisely sized DNA fragments ranging from 100bp to 20kb. This ready-to-use ladder features enhanced intensity reference bands at 1kb, 3kb, 6kb, and 20kb positions for simplified molecular weight determination.
Catalog No. NC3726411 Supplier LAMDA BIOTECH INC Supplier No. M10820
The 1Kb Plus™ DNA Ladder-20 is a comprehensive molecular weight marker containing 14 precisely sized DNA fragments ranging from 100bp to 20kb. This ready-to-use ladder features enhanced intensity reference bands at 1kb, 3kb, 6kb, and 20kb positions for simplified molecular weight determination.