Learn More
LAMDA BIOTECH INC 1Kb Quant Logic DNA Ladder
Catalog No. NC3726047
Encompass_Preferred
A precision-designed molecular weight marker featuring 16 DNA fragments from 100bp to 10Kb. Each band contains a proportional DNA mass relative to its size, enabling straightforward DNA fragment quantification during gel electrophoresis.
Catalog No. NC3726047 Supplier LAMDA BIOTECH INC Supplier No. M103
A precision-designed molecular weight marker featuring 16 DNA fragments from 100bp to 10Kb. Each band contains a proportional DNA mass relative to its size, enabling straightforward DNA fragment quantification during gel electrophoresis.