Learn More
Ace Glass, Inc. Flask, recovery, heavy wall, 1L, 24/40
SDP
Catalog No. 509557460
Encompass_Preferred
1L round bottom heavy wall recovery flask, with a single neck with a 24/40 joint, and a sloping side wall.
Catalog No. 50-955-7460 Supplier Ace Glass, Inc. Supplier No. 6892230
1L round bottom heavy wall recovery flask, with a single neck with a 24/40 joint, and a sloping side wall.