Learn More
Chemglass Life Sciences 1000mL Addition Funnel, 24/40 Joint Size, 4mm Glass Stpk, 520mm OAH
SDP
Catalog No. 501531213
Encompass_Preferred
1000mL Addition Funnel, 24/40 Joint Size, 4mm Glass Stpk, 520mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. 50-153-1213 Supplier Chemglass Life Sciences Supplier No. CG170006
1000mL Addition Funnel, 24/40 Joint Size, 4mm Glass Stpk, 520mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.