Learn More
Chemglass Life Sciences 1000mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Metering Stpk, 535mm OAH
SDP
Catalog No. CG171321
Encompass_Preferred
1000mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Metering Stopcock, 535mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171321 Supplier Chemglass Life Sciences Supplier No. CG171321
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
1000mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Metering Stopcock, 535mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.