Learn More
FUJIFILM Biosciences BalanCD Transfectory CHO - Liquid
Catalog No. NC1348122
Encompass
BalanCD Transfectory CHO system of media and supplements is a scalable platform that produces milligram to gram-scale yields of recombinant proteins using a simplified, affordable protocol. The system (including BalanCD Transfectory CHO, Transfectory Supplement and Anti-Clumping Supplement) supports small- andLarge-scale transient transfection in CHO cells, enabling scientists to use the same medium before and after transfection.The chemically-defined, animal component-free medium produces proteinLevels equivalent to, or better than, other commercially available media.
Catalog No. NC1348122 Supplier FUJIFILM Biosciences Supplier No. 911471L
BalanCD Transfectory CHO system of media and supplements is a scalable platform that produces milligram to gram-scale yields of recombinant proteins using a simplified, affordable protocol. The system (including BalanCD Transfectory CHO, Transfectory Supplement and Anti-Clumping Supplement) supports small- andLarge-scale transient transfection in CHO cells, enabling scientists to use the same medium before and after transfection.The chemically-defined, animal component-free medium produces proteinLevels equivalent to, or better than, other commercially available media.