Learn More
Cole-Parmer Glass Dewar Flask with Handle, Lid, and Lid Clamps, Blue Enameled Steel, 1 L
Catalog No. NC1264358 Shop All Cole-Parmer Products
Encompass_Preferred
Blue Enameled Steel and Glass Dewar Flask with Handle, Lid, and Lid Clamps, 1 L, Safely transport liquid gasesIdeal for use with cold trapsCushioned base prevents benchtop damageInsulated lid and fold-down handle make it easy to transport
Catalog No. NC1264358 Supplier Cole-Parmer Supplier No. 0377422
Blue Enameled Steel and Glass Dewar Flask with Handle, Lid, and Lid Clamps, 1 L, Safely transport liquid gasesIdeal for use with cold trapsCushioned base prevents benchtop damageInsulated lid and fold-down handle make it easy to transport