Learn More
HCL Labels 1L Pre-labeled GHS Compliant Dual-Action Kwazar Spray Bottle Distilled Water, Bottle Supplied Empty, 1EA
SDP
Catalog No. 501633568
Encompass_Preferred
Bottles, HCL Labels, Inc., 1L, GHS Distilled Water Spray Bottle, These Kwazar bottles are pre-labeled with a GHS hazard communication chemical label. Bottles are chemical resistant and empty.
Catalog No. 50-163-3568 Supplier HCL Labels Supplier No. GHSKWBOT0025
Bottles, HCL Labels, Inc., 1L, GHS Distilled Water Spray Bottle, These Kwazar bottles are pre-labeled with a GHS hazard communication chemical label. Bottles are chemical resistant and empty.