Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1L-epi-2-Inosose 98.0+%, TCI America™
SDP

Catalog No. I0634200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0634200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0634200MG Supplier TCI America Supplier No. I0634200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33471-33-9
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00038395
InChI Key VYEGBDHSGHXOGT-SHRIJZPSSA-N
Synonym 2L-2,3,4,6/5-Pentahydroxycyclohexanone
PubChem CID 9877530
IUPAC Name (2S,3S,5S,6R)-2,3,4,5,6-pentahydroxycyclohexan-1-one
SMILES C1(C(C(C(=O)C(C1O)O)O)O)O

Specifications

Color White
Quantity 200 mg
Formula Weight 178.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1L-epi-2-Inosose
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.