Learn More
Chemglass Life Sciences 1000mL Erlenmeyer Flask, 38-430 GPI Screw Thread
SDP
Catalog No. CG154305
Encompass_Preferred
1000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. CG154305 Supplier Chemglass Life Sciences Supplier No. CG154305
1000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.