Learn More
Ace Glass, Inc. 1L FLASK 1/4 HC C 1L FLASK 1/4 HC C
SDP
Supplier:  Ace Glass, Inc. 698351
Encompass_Preferred
1L heavy wall filter flask with a tooled neck for uniform stopper fit. Side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. Includes flask, 1/4¿ tubing connector, and nylon bushing.
Catalog No. 50-973-980