Learn More
Chemglass Life Sciences 1, 000mL Erlenmeyer Flask
SDP
Catalog No. 501215019
Encompass_Preferred
1,000mL Erlenmeyer Flask. Tops are reinforced and tooled with a rounded finish, body is thick walled with a long tapered outside contour to minimize chipping when struck or rubbed together. All flasks have durable white ceramic enamel scales to indicate approximate volumes. Designed from ASTM Specification E1404, Type I, Class 1 requirements.
Catalog No. 50-121-5019 Supplier Chemglass Life Sciences Supplier No. CG8496P1L
1,000mL Erlenmeyer Flask. Tops are reinforced and tooled with a rounded finish, body is thick walled with a long tapered outside contour to minimize chipping when struck or rubbed together. All flasks have durable white ceramic enamel scales to indicate approximate volumes. Designed from ASTM Specification E1404, Type I, Class 1 requirements.