Learn More
Chemglass Life Sciences 1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Stpk, 525mm OAH
SDP
Catalog No. CG171206
Encompass_Preferred
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Stopcock, 525mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171206 Supplier Chemglass Life Sciences Supplier No. CG171206
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm PTFE Stopcock, 525mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.