Learn More
Chemglass Life Sciences 1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm Glass Stpk, 525mm OAH
SDP
Catalog No. CG171006
Encompass_Preferred
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm Glass Stopcock, 525mm OAH. Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171006 Supplier Chemglass Life Sciences Supplier No. CG171006
1000mL Addition Funnel, Graduated, 24/40 Joint Size, 4mm Glass Stopcock, 525mm OAH. Cylindrical style addition funnel graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.