Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0785131
Encompass_Preferred
Fabric heating mantle for 1L Griffin beaker to 400 degrees C maximum internal operating temperature. 430W, 115V, CSA.
Catalog No. NC0785131 Supplier Glas Col LLC Supplier No. 100A O614
Fabric heating mantle for 1L Griffin beaker to 400 degrees C maximum internal operating temperature. 430W, 115V, CSA.