Learn More
Ace Glass, Inc. 1000mL jacketed filter reactor, complete with 28/15 ballpoint inlet/outlets, 100mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings
SDP
Catalog No. 50972803
Encompass
1000mL jacketed filter reactor, complete with 28/15 ballpoint inlet/outlets, 100mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings
Catalog No. 50-972-803 Supplier Ace Glass, Inc. Supplier No. 638650
1000mL jacketed filter reactor, complete with 28/15 ballpoint inlet/outlets, 100mm four neck 24/40 head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, clamp, bushings & O-rings