Learn More
Ace Glass, Inc. 1L heavy wall filter reactor with five neck head, #80threaded body, 10mm stir shaft and bearing, and 3/8" female NPT 100 micron polyethylene filter adapter. Pressure rated at 35 psi.
SDP
Catalog No. 50972796
Encompass
Catalog No. 50-972-796 Supplier Ace Glass, Inc. Supplier No. 6384235