Learn More
Chemglass Life Sciences 1000mL Single Neck Round Bottom Flask (RBF) , 24/40 Outer Joint, Thermowell
SDP
Catalog No. CG151505
Encompass_Preferred
1000mL Single Neck Round Bottomed Flask, 24/40 Outer Joint, Thermowell. Round bottom, HEAVY WALL flask with a standard taper outer joint and a side well to accept a thermometer. Top of thermowell is tooled to accept a #0 rubber stopper.
Catalog No. CG151505 Supplier Chemglass Life Sciences Supplier No. CG151505
1000mL Single Neck Round Bottomed Flask, 24/40 Outer Joint, Thermowell. Round bottom, HEAVY WALL flask with a standard taper outer joint and a side well to accept a thermometer. Top of thermowell is tooled to accept a #0 rubber stopper.