Learn More
Ace Glass, Inc. 1L Instatherm filter reactor body with 100mm ID grooved top flange, #80 bottom thread, 100mm IDx180mm depth, with CAPFE O-ring and power cord, 120v/5amp max
SDP
Catalog No. 50972821
Encompass
1L Instatherm coated filter reactor body with a 100mm I.D. grooved top flange for connection to a reaction head with the use of a quick release clamp, and a #80 Ace-Thred bottom thread for use with a bottom adapter with a filter support and drain valve. Instatherm provides safer, rapid heating, and can be operated at 115v with ACE Proportional Temperature Controllers, or with a Powerstat. Reactor body supplied with a CAPFE o-ring and a power cord. Dimensions: 100mm I.D. x 180mm D.
Catalog No. 50-972-821 Supplier Ace Glass, Inc. Supplier No. 638825
1L Instatherm coated filter reactor body with a 100mm I.D. grooved top flange for connection to a reaction head with the use of a quick release clamp, and a #80 Ace-Thred bottom thread for use with a bottom adapter with a filter support and drain valve. Instatherm provides safer, rapid heating, and can be operated at 115v with ACE Proportional Temperature Controllers, or with a Powerstat. Reactor body supplied with a CAPFE o-ring and a power cord. Dimensions: 100mm I.D. x 180mm D.