Learn More
Ace Glass, Inc. 1000mL jacketed pressure reactor, complete with 28/15 ballpoint I/O, 100mm Ace-Thred five neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
SDP
Catalog No. 50978675
Encompass
1000mL jacketed pressure reactor, complete with 28/15 ballpoint I/O, 100mm Ace-Thred five neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
Catalog No. 50-978-675 Supplier Ace Glass, Inc. Supplier No. 6386115
1000mL jacketed pressure reactor, complete with 28/15 ballpoint I/O, 100mm Ace-Thred five neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings