Learn More
Ace Glass, Inc. 1000mL separatory funnel with locking stopper
SDP
Catalog No. 50974701
Encompass_Preferred
1000ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-701 Supplier Ace Glass, Inc. Supplier No. 723163
1000ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.