Learn More
S2 Media 1L Mueller Hinton (MH) Broth, 1L PET/PETG Media Bottle, 1/each
Catalog No. NC3900352
Encompass_Preferred
Mueller Hinton (MH) Broth is a nonselective, nondifferential nutrient-rich liquid medium that supports the growth of a wide range of non-fastidious bacteria. It is primarily intended for standardized antimicrobial susceptibility testing, such as determining minimum inhibitory concentrations (MICs), by providing a consistent, interference-free environment for bacterial growth. This medium is not supplemented with calcium or magnesium ions.• Product meets ASM and CLSI reference methods and performance criteria.• MH Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• This formulation is not cation adjusted.
Catalog No. NC3900352 Supplier S2 Media Supplier No. 5525
Mueller Hinton (MH) Broth is a nonselective, nondifferential nutrient-rich liquid medium that supports the growth of a wide range of non-fastidious bacteria. It is primarily intended for standardized antimicrobial susceptibility testing, such as determining minimum inhibitory concentrations (MICs), by providing a consistent, interference-free environment for bacterial growth. This medium is not supplemented with calcium or magnesium ions.• Product meets ASM and CLSI reference methods and performance criteria.• MH Broth should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• This formulation is not cation adjusted.