Learn More
Ace Glass, Inc. 1000mL separatory funnel body only, pear shaped, #11 Ace-Thred bottom, includes 4mm bore PTFE stopcock plug, 7223-40 #11 PTFE drip tip & 12632-27 size 27 PTFE stopper not included
SDP
Catalog No. 50974665
Encompass_Preferred
1000ml pear-shaped, separatory funnel body. Funnel has a 4mm bore PTFE stopcock plug and a #11 Ace-Thred bottom which creates a leak tight connection without the need for an o-ring.
Catalog No. 50-974-665 Supplier Ace Glass, Inc. Supplier No. 722320
1000ml pear-shaped, separatory funnel body. Funnel has a 4mm bore PTFE stopcock plug and a #11 Ace-Thred bottom which creates a leak tight connection without the need for an o-ring.