Learn More
Ace Glass, Inc. 1000mL separatory funnel, pear shaped, complete with #11 removable PTFE drip tip and 4mm bore PTFE stopcock plug, 12632-27 size 27 PTFE stopper not included
SDP
Catalog No. 50974672
Encompass_Preferred
1000ml pear-shaped, separatory funnel complete with #11 removable PTFE drip tip and 4mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.
Catalog No. 50-974-672 Supplier Ace Glass, Inc. Supplier No. 722359
1000ml pear-shaped, separatory funnel complete with #11 removable PTFE drip tip and 4mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.