Learn More
Alkali Scientific 1L PETG Square Storage Bottle
SDP
Supplier:  Alkali Scientific EFC826
Encompass_Preferred
The Liquid Transfer System collection provides an aseptic transfer solution of closed liquid infusion for the pharmaceuticals and biotechnology industry as well as laboratories A standard Luer head is used in the joint of the system enabling the operation easier and more convenient Also the TPE material of tubes is in line with the requirements for industrial pharmaceutical enterprises and is validated by a certified third party in accordance with BPOG Guide for Evaluating Extractables and Leachables This Square Storage Bottle Closed System Cap is an ideal choice for laboratories and industries requiring reliable sterile and durable liquid transfer and storage solutions They are lot tested and certified free of endotoxins and pyrogens Complete piping specifications Transfer caps with integrated injection molding wide versatility can be adapted to other brands High chemical durability suitable for the storage of high purity reagent standard particle materials and samples Highly
Catalog No. 50-302-7592