Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 1000mL round bottom flasks, max diameter 5.12 inches, 240v 290 watts per position, order code (-63) or (-73) power supply separately
SDP
Catalog No. 50977367
Encompass
Combo mantle, 3 position, low temperature, fits 1000mL round bottom flasks, max diameter 5.12 inches, 240v 290 watts per position, order code (-63) or (-73) power supply separately
Catalog No. 50-977-367 Supplier Ace Glass, Inc. Supplier No. 12061140
Combo mantle, 3 position, low temperature, fits 1000mL round bottom flasks, max diameter 5.12 inches, 240v 290 watts per position, order code (-63) or (-73) power supply separately