Learn More
Alkali Scientific 1L Roller Bottles 1000mL 490cm
SDP
Catalog No. 503028170
Encompass_Preferred
Our 1000mL Roller Bottle is designed for tissue culture applications It features a length of 49 cm and is tissue culture treated for optimal cell adhesion and growth The bottle is equipped with printed graduations for easy volume reading The non-vented cap helps maintain a sterile environment within the bottle to prevent contamination Each unit is sterilized ensuring aseptic conditions for your experiments Ideal for cell culture work in research laboratories pharmaceutical or biotechnology industries this roller bottle provides a convenient vessel for cell growth and propagation Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged sterile in individual bags Vent caps feature 0
Catalog No. 50-302-8170 Supplier Alkali Scientific Supplier No. VT1000P
Our 1000mL Roller Bottle is designed for tissue culture applications It features a length of 49 cm and is tissue culture treated for optimal cell adhesion and growth The bottle is equipped with printed graduations for easy volume reading The non-vented cap helps maintain a sterile environment within the bottle to prevent contamination Each unit is sterilized ensuring aseptic conditions for your experiments Ideal for cell culture work in research laboratories pharmaceutical or biotechnology industries this roller bottle provides a convenient vessel for cell growth and propagation Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged sterile in individual bags Vent caps feature 0