Learn More
Accuflow Systems Saint-Gobain Media Bottle Assembly, 1 L, PETG, C-Flex Tubing
Catalog No. NC2312804
Encompass_Preferred
Bio-Simplex media bottle assembly is a sterile 1 L PETG bottle system with a two-port closure, integrated biopharmaceutical TPE tubing, and a 0.2 μm vent filter. The assembly is gamma irradiated and double-bagged for single-use bioprocess applications and is compatible with sterile welding and thermal tubing sealers.
  • Capacity 1 L (1000 mL)
  • Bottle material PETG
  • Closure Two-port integral closure with thermal bond to tubing
  • Tubing 18 in (46 cm), 1/8 in (3.2 mm) I.D., 1/4 in (6.4 mm) O.D., plug end
  • Vent filter 0.2 μm, 25 mm diameter
  • Sterilization Gamma irradiated to SAL 10^-6; sold double-bagged
  • Applications Single-use bioprocessing, aseptic media handling, sterile connections
Catalog No. NC2312804 Supplier Accuflow Systems Supplier No. JSA38MM1L2FDB
Bio-Simplex media bottle assembly is a sterile 1 L PETG bottle system with a two-port closure, integrated biopharmaceutical TPE tubing, and a 0.2 μm vent filter. The assembly is gamma irradiated and double-bagged for single-use bioprocess applications and is compatible with sterile welding and thermal tubing sealers.
  • Capacity 1 L (1000 mL)
  • Bottle material PETG
  • Closure Two-port integral closure with thermal bond to tubing
  • Tubing 18 in (46 cm), 1/8 in (3.2 mm) I.D., 1/4 in (6.4 mm) O.D., plug end
  • Vent filter 0.2 μm, 25 mm diameter
  • Sterilization Gamma irradiated to SAL 10^-6; sold double-bagged
  • Applications Single-use bioprocessing, aseptic media handling, sterile connections