Learn More
S2 Media 1L Synthetic Broth, AOAC, 1L PET Media Bottle, 1/each
Catalog No. NC3900349
Encompass_Preferred
Synthetic Broth, AOAC is a selective, non-differential, chemically defined medium formulated for the maintenance and propagation of disinfectant test cultures. It supplies all essential nutrients required to sustain the growth of standardized test organisms employed in the determination of phenol coefficients for disinfectant efficacy evaluation.• Also known as Mundy and Wright Broth• Product meets AOAC reference methods and performance criteria.• Synthetic Broth, AOAC should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. NC3900349 Supplier S2 Media Supplier No. 5523
Synthetic Broth, AOAC is a selective, non-differential, chemically defined medium formulated for the maintenance and propagation of disinfectant test cultures. It supplies all essential nutrients required to sustain the growth of standardized test organisms employed in the determination of phenol coefficients for disinfectant efficacy evaluation.• Also known as Mundy and Wright Broth• Product meets AOAC reference methods and performance criteria.• Synthetic Broth, AOAC should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.