Learn More
Chemglass Life Sciences Chemglass Life Sciences 1 L Unjacketed Bioreactor Vessel (CLS-1378-01)
SDP
Catalog No. 5000083295
Encompass
The Chemglass Life Sciences one liter unjacketed bioreactor vessel is a glass containment component for laboratory-scale microbial fermentation and cell culture systems. It is supplied as the standalone vessel and is designed to integrate with compatible headplates, agitators, and support accessories to form a complete one liter bioreactor setup.
  • One liter nominal volume.
  • Unjacketed design for direct use without a temperature jacket.
  • Glass construction for chemical resistance and visual monitoring.
  • Compatible with headplate assemblies that provide multiple ports.
  • Vessel only; headplate, motor, and accessories sold separately.
Catalog No. 50-000-83295 Supplier Chemglass Life Sciences Supplier No. CLS137801
The Chemglass Life Sciences one liter unjacketed bioreactor vessel is a glass containment component for laboratory-scale microbial fermentation and cell culture systems. It is supplied as the standalone vessel and is designed to integrate with compatible headplates, agitators, and support accessories to form a complete one liter bioreactor setup.
  • One liter nominal volume.
  • Unjacketed design for direct use without a temperature jacket.
  • Glass construction for chemical resistance and visual monitoring.
  • Compatible with headplate assemblies that provide multiple ports.
  • Vessel only; headplate, motor, and accessories sold separately.