Learn More
Chemglass Life Sciences Chemglass 125 mm Watch Glass, Fits 1 L Beaker
SDP
Catalog No. NC2217092
Encompass_Preferred
This 125 mm watch glass is a concave borosilicate laboratory glass cover and evaporating surface sized to fit a 1,000 mL (1 L) beaker. It provides a durable, stackable surface for evaporation, weighing, and beaker coverage.
  • 125 mm diameter; fits a 1,000 mL beaker.
  • Borosilicate glass construction for chemical durability.
  • Heavy wall with fire-polished edge for mechanical strength.
  • Molded radius of curvature for convenient stacking.
  • Outside diameter may vary slightly; slight out-of-round tolerance.
  • Suitable as a beaker cover, evaporating dish, or sample holder.
This item is not returnable. View return policy
Catalog No. NC2217092 Supplier Chemglass Life Sciences Supplier No. CG8242500
This 125 mm watch glass is a concave borosilicate laboratory glass cover and evaporating surface sized to fit a 1,000 mL (1 L) beaker. It provides a durable, stackable surface for evaporation, weighing, and beaker coverage.
  • 125 mm diameter; fits a 1,000 mL beaker.
  • Borosilicate glass construction for chemical durability.
  • Heavy wall with fire-polished edge for mechanical strength.
  • Molded radius of curvature for convenient stacking.
  • Outside diameter may vary slightly; slight out-of-round tolerance.
  • Suitable as a beaker cover, evaporating dish, or sample holder.