Learn More
Andwin Scientific 1LB 3MM GLASS BEAD
SDP
Catalog No. NC9385575
Encompass_Preferred
The glass beads are constructed with soda lime glass with a gradvity of 2.49. This product is sold in a quantity of 1 pound.
Catalog No. NC9385575 Supplier Andwin Scientific Supplier No. 5663L13
The glass beads are constructed with soda lime glass with a gradvity of 2.49. This product is sold in a quantity of 1 pound.