Learn More
Cayman Chemical 1Linoleoyl Glycerol
SDP
Catalog No. 502889013
Encompass_Preferred
A fatty acid glycerol that has been isolated from S. chinensis roots, inhibits PAF-AH with an IC50 value of 45 µM
Catalog No. 50-288-9013 Supplier Cayman Chemical Supplier No. 1000886925 MG
Add to Cart
Add to Cart
A fatty acid glycerol that has been isolated from S. chinensis roots, inhibits PAF-AH with an IC50 value of 45 µM