Learn More
Cayman Chemical 1-Linoleoyl Glycerol, Purity: ≥90%, CAS: 2277-28-3, 10 mg
SDP
Catalog No. 502890157
Encompass_Preferred
A fatty acid glycerol that has been isolated from S. chinensis roots, inhibits PAF-AH with an IC50 value of 45 µM
Catalog No. 50-289-0157 Supplier Cayman Chemical Supplier No. 1000886910 MG
A fatty acid glycerol that has been isolated from S. chinensis roots, inhibits PAF-AH with an IC50 value of 45 µM