Learn More
Cayman Chemical 1Linoleoyl Glycerol
SDP
Supplier:  Cayman Chemical 100088691 MG
Encompass_Preferred
A fatty acid glycerol that has been isolated from S. chinensis roots, inhibits PAF-AH with an IC50 value of 45 µM
Catalog No. 50-288-8721