Learn More
Cayman Chemical 1LInoleoyl3aLInolenoylra 10mg
SDP
Catalog No. 502935227
Encompass_Preferred
A diacylglycerol; has been found in olive oil subjected to lipase-catalyzed glycerolysis with immobilized lipase B from C. antarctica
Catalog No. 50-293-5227 Supplier Cayman Chemical Supplier No. 2682910 MG
A diacylglycerol; has been found in olive oil subjected to lipase-catalyzed glycerolysis with immobilized lipase B from C. antarctica