Learn More
Cayman Chemical 1LInoleoyl3PalmItoylracg 10mg
SDP
Supplier:  Cayman Chemical 2817610 MG
Encompass_Preferred
A diacylglycerol; has been found in wheat bran extracts and dry-cured, but not raw, Iberian ham
Catalog No. 50-293-9227