Learn More
Cayman Chemical 1LInoleoyld112LInoleoyls 100UG
SDP
Catalog No. 502896119
Encompass_Preferred
An internal standard for the quantification of 1,2-dilinoleoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-289-6119 Supplier Cayman Chemical Supplier No. 9004419100 UG
An internal standard for the quantification of 1,2-dilinoleoyl-sn-glycero-3-PC by GC- or LC-MS