Learn More
Chemglass Life Sciences 1000mL Addition Funnel, 24/40 Joint Size, 4mm PTFE Stpk, 520mm OAH
SDP
Catalog No. CG170206
Encompass_Preferred
1000mL Addition Funnel, 24/40 Joint Size, 4mm PTFE Stopcock, 520mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.
Catalog No. CG170206 Supplier Chemglass Life Sciences Supplier No. CG170206
1000mL Addition Funnel, 24/40 Joint Size, 4mm PTFE Stopcock, 520mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.