Learn More
Foxx Life Sciences Foxx Life Sciences Heavy Duty Vacuum Bottles
GREENER_CHOICE
Catalog No. NC2128278
Encompass
Foxx Life Sciences Heavy Duty Vacuum Bottles have thick walls that can hold near full vacuum for 24 hours, making them excellent replacements for glass HPLC containers, and ideal for applications involving liquid aspiration, cell harvesting, and degasification. All Heavy Duty Vacuum Bottles are compatible with either 53B or 83B VersaCap technology and adapters, enabling easy customization of the bottle to fit your application.
This item is not returnable. View return policy
Catalog No. NC2128278 Supplier Foxx Life Sciences Supplier No. 1621102OEM
Foxx Life Sciences Heavy Duty Vacuum Bottles have thick walls that can hold near full vacuum for 24 hours, making them excellent replacements for glass HPLC containers, and ideal for applications involving liquid aspiration, cell harvesting, and degasification. All Heavy Duty Vacuum Bottles are compatible with either 53B or 83B VersaCap technology and adapters, enabling easy customization of the bottle to fit your application.