Learn More
Genesee Scientific Corporation Olympus 1.7mL Snap Cap Microcentrifuge Tubes, Positive Seal Design, RNase/DNase Free, Sterile
SDP
Catalog No. NC2145897
Encompass
Olympus 1.7mL Microcentrifuge Tubes offer exceptional clarity and durability for laboratory applications. With a positive snap cap design, they prevent sample loss during boiling and centrifugation. These versatile tubes are certified RNase, DNase, and PCR inhibitor-free, ensuring integrity for sensitive workflows. Autoclavable and boil-proof, they withstand extreme temperatures, making them ideal for a wide range of experiments.
This item is not returnable. View return policy
Catalog No. NC2145897 Supplier Genesee Scientific Corporation Supplier No. 22281S
Olympus 1.7mL Microcentrifuge Tubes offer exceptional clarity and durability for laboratory applications. With a positive snap cap design, they prevent sample loss during boiling and centrifugation. These versatile tubes are certified RNase, DNase, and PCR inhibitor-free, ensuring integrity for sensitive workflows. Autoclavable and boil-proof, they withstand extreme temperatures, making them ideal for a wide range of experiments.