Learn More
Electron Microscopy Sciences Poly-L-Lysine German Glass Coverslips #1, 25 mm
SDP
Catalog No. NC1539821
Encompass_Preferred
CE certified crystal clear German glass coverslips sterilized by radiation and ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 25 mm
Catalog No. NC1539821 Supplier Electron Microscopy Sciences Supplier No. 7229219
CE certified crystal clear German glass coverslips sterilized by radiation and ready to use. Tested for cell and neuron cultures. User manual included.
Diameter: 25 mm