Learn More
Teknova 1M Arginine Solution, pH 8.5. 1000mL, Sterile
SDP
Supplier:  Teknova A8330
Encompass
Amino Acids are essential nutrients for cell growth, and thus they are often used in the preparation of growth media. Amino acid solution can help to increase cell viability and growth rate, making it an essential component for cell culture.
Catalog No. 50-238-8159