Learn More
Teknova 1M Citric Acid, pH 2.95. 1000mL, Sterile 2 pack.
SDP
Catalog No. 502387264
Encompass
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.
Catalog No. 50-238-7264 Supplier Teknova Supplier No. C24962PK
Sodium citrate solution is a commonly used reagent in life science. It has many uses, such as making media for the growth of microorganisms, lysing cells, and precipitating nucleic acids.