Learn More
GFS Chemicals Inc CUPRIETHYLENEDIAMINE SOLUTION, 1M | 4L | GFS CHEMICALS, INC. Item #: 32101 | Bulk #: 666
SDP
Catalog No. NC9874290
Encompass_Preferred
CUPRIETHYLENEDIAMINE SOLUTION, 1M | Chemical Formula : Cu(C!2H!8N!2)!2(OH)!2 | CAS # : 14552-35-3 | Specifications : Ethylenediamine to copper ratio 1.96 - 2.04
Catalog No. NC9874290 Supplier GFS Chemicals Inc Supplier No. 32101
CUPRIETHYLENEDIAMINE SOLUTION, 1M | Chemical Formula : Cu(C!2H!8N!2)!2(OH)!2 | CAS # : 14552-35-3 | Specifications : Ethylenediamine to copper ratio 1.96 - 2.04