Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc HEPES Buffer (1 M), pH 7.3, 100 mL
SDP

Catalog No. 507517290
Encompass_Preferred

HEPES is a zwitterionic organic chemical buffering agent. HEPES (1M) is used as a buffer alternative from sodium bicarbonate for cell culture and is widely used in many biochemical reactions and as a buffering agent in some cell culture media. HEPES is reportedly superior to NaHCO3 in controlling pH in tissue and organ culture.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Performance Tested
pH: 7.2-7.4
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7290 Supplier Quality Biological Inc Supplier No. 118089721EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HEPES is a zwitterionic organic chemical buffering agent. HEPES (1M) is used as a buffer alternative from sodium bicarbonate for cell culture and is widely used in many biochemical reactions and as a buffering agent in some cell culture media. HEPES is reportedly superior to NaHCO3 in controlling pH in tissue and organ culture.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Performance Tested
pH: 7.2-7.4
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.